missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine ethyl ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7054417
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14719-37-0
C9H17NO4
203.238
MFCD05663720
CNIBHMMDDXGDNR-UHFFFAOYSA-N
n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
357729
ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
CCOC(=O)CNC(=O)OC(C)(C)C
This item is not returnable. View return policy
14719-37-0
C9H17NO4
203.238
MFCD05663720
CNIBHMMDDXGDNR-UHFFFAOYSA-N
n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
357729
ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
CCOC(=O)CNC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Linear Formula (CH3)3COCONHCH2CO2CH2CH3
Formula Weight 203.24
Percent Purity 95%
Quantity 5 g
Physical Form Oily Liquid
Chemical Name or Material N-Boc-glycine ethyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'