missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-valinol, 95%, Thermo Scientific™

Catalog Number p-7053600
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
106391-87-1
C10H21NO3
203.28
MFCD00235960
OOQRRYDVICNJGC-QMMMGPOBSA-N
boc-d-valinol, n-boc-d-valinol, r-tert-butyl 1-hydroxy-3-methylbutan-2-yl carbamate, tert-butyl n-2r-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, tert-butyl n-1r-1-hydroxymethyl-2-methyl-propyl carbamate, n-tert-butoxycarbonyl-d-valinol, n-t-boc-d-valinol, ksc491q4r
11241137
tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
CC(C)[C@H](CO)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
106391-87-1
C10H21NO3
203.28
MFCD00235960
OOQRRYDVICNJGC-QMMMGPOBSA-N
boc-d-valinol, n-boc-d-valinol, r-tert-butyl 1-hydroxy-3-methylbutan-2-yl carbamate, tert-butyl n-2r-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, tert-butyl n-1r-1-hydroxymethyl-2-methyl-propyl carbamate, n-tert-butoxycarbonyl-d-valinol, n-t-boc-d-valinol, ksc491q4r
11241137
tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
CC(C)[C@H](CO)NC(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 218°C
Color Yellow
Refractive Index 1.451
Formula Weight 203.28
Percent Purity 95%
Density 0.995 g/mL
Quantity 1 g
Physical Form Liquid
Chemical Name or Material N-Boc-D-valinol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'