missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid 1-benzyl ester, 99%, Thermo Scientific Chemicals

Catalog Number p-7026125
40.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
34404-30-3
C17H23NO6
337.372
MFCD00038266
CVZUKWBYQQYBTF-CYBMUJFWSA-N
boc-d-glu-obzl, boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid benzyl ester, 1-benzyl n-boc-l-glutamate, n-tert-butoxycarbonyl-d-glutamic acid 1-benzyl ester, 4r-5-benzyloxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-obn, pubchem12942, 1-benzyl n-boc-d-glutamate
7010515
(4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
34404-30-3
C17H23NO6
337.372
MFCD00038266
CVZUKWBYQQYBTF-CYBMUJFWSA-N
boc-d-glu-obzl, boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid benzyl ester, 1-benzyl n-boc-l-glutamate, n-tert-butoxycarbonyl-d-glutamic acid 1-benzyl ester, 4r-5-benzyloxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-obn, pubchem12942, 1-benzyl n-boc-d-glutamate
7010515
(4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 337.37
Percent Purity 99%
Chemical Name or Material N-Boc-D-glutamic acid 1-benzyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'