missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientific™

Catalog Number p-7041597
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
112898-18-7
C15H25NO6
315.37
MFCD00060741
NLPQIWFEEKQBBN-UHFFFAOYNA-N
boc-d-asp ochex-oh, boc-d-aspartic acid 4-cyslohexyl ester, boc-d-asp ochx-oh, r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, 2r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-d-asp ochx-oh tlc, n-alpha-t-boc-d-aspartic beta-cyclohexyl ester, n-tert-butyloxycarbonyl-d-aspartic acid 4-cyslohexyl ester
7016592
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(O)=O
This item is not returnable. View return policy
112898-18-7
C15H25NO6
315.37
MFCD00060741
NLPQIWFEEKQBBN-UHFFFAOYNA-N
boc-d-asp ochex-oh, boc-d-aspartic acid 4-cyslohexyl ester, boc-d-asp ochx-oh, r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, 2r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-d-asp ochx-oh tlc, n-alpha-t-boc-d-aspartic beta-cyclohexyl ester, n-tert-butyloxycarbonyl-d-aspartic acid 4-cyslohexyl ester
7016592
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-4-(cyclohexyloxy)-4-oxobutanoic acid
Formula Weight 315.36
Percent Purity 95%
Chemical Name or Material N-Boc-D-aspartic acid 4-cyclohexyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'