missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-D-proline methyl ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7054738
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
114676-69-6
C11H19NO5
245.275
MFCD00797548
MZMNEDXVUJLQAF-HTQZYQBOSA-N
2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome
6951202
1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O
This item is not returnable. View return policy
114676-69-6
C11H19NO5
245.275
MFCD00797548
MZMNEDXVUJLQAF-HTQZYQBOSA-N
2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome
6951202
1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 245.27
Percent Purity 95%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Boc-cis-4-hydroxy-D-proline methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'