missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-oxo-L-proline tert-butyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7029862
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
166410-05-5
C14H23NO5
285.34
MFCD06797013
MPNWQUWKRDADHK-JTQLQIEISA-N
n-boc-4-oxo-l-proline tert-butyl ester, s-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-proline tert-butyl ester, 1,2-di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-prolinetert-butylester, di-tert-butyl s-4-oxopyrrolidine-1,2-dicarboxylate, di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butoxycarbonyl-4-oxo-l-proline tert-butyl ester, 1-tert-butyloxycarbonyl-4-oxo-l-proline tert-butyl ester, s-4-oxopyrrolidine-1,2-dicarboxylic acid di-tert-butyl ester
11346670
ditert-butyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C
This item is not returnable. View return policy
166410-05-5
C14H23NO5
285.34
MFCD06797013
MPNWQUWKRDADHK-JTQLQIEISA-N
n-boc-4-oxo-l-proline tert-butyl ester, s-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-proline tert-butyl ester, 1,2-di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-prolinetert-butylester, di-tert-butyl s-4-oxopyrrolidine-1,2-dicarboxylate, di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butoxycarbonyl-4-oxo-l-proline tert-butyl ester, 1-tert-butyloxycarbonyl-4-oxo-l-proline tert-butyl ester, s-4-oxopyrrolidine-1,2-dicarboxylic acid di-tert-butyl ester
11346670
ditert-butyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 64°C to 67°C
Quantity 250 mg
Formula Weight 285.35
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'