missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2,4,5-trifluoro-D-phenylalanine, 98%, Thermo Scientific™

Catalog Number p-7054566
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
486460-09-7
C14H16F3NO4
319.28
MFCD06659118
FTWHJNNFVISGNQ-LLVKDONJSA-N
r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-2,4,5-trifluoro-d-phenylalanine, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-2,4,5-trifluorophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-d-2-amino-3-2,4,5-trifluoro-phenyl alanine, r-n-tert-butoxycarbonyl-2,4,5-trifluorophenylalanine, r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoicacid
7146200
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(2,4,5-trifluorophenyl)propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O
This item is not returnable. View return policy
486460-09-7
C14H16F3NO4
319.28
MFCD06659118
FTWHJNNFVISGNQ-LLVKDONJSA-N
r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-2,4,5-trifluoro-d-phenylalanine, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-2,4,5-trifluorophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-d-2-amino-3-2,4,5-trifluoro-phenyl alanine, r-n-tert-butoxycarbonyl-2,4,5-trifluorophenylalanine, r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoicacid
7146200
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(2,4,5-trifluorophenyl)propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 319.28
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Boc-2,4,5-trifluoro-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'