Learn More
MP Biomedicals™ b-Lactamase [E.C. 3.5.2.6]
From Bacillus cereus
Brand:  MP Biomedicals™ 0219003401
Additional Details : CAS : 9073-60-3 Weight : 0.00100kg
Product Code. 11474472
  • 532.12 € / Each
Estimated Shipment: 07-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Lyophilized solid. Activity: Each vial contains 500U β-Lactamase I and 50U β-Lactamase II. Unit Definition: One unit is defined as the amount of enzyme which will hydrolyze 1.0μm of benzyl penicillin and 1.0μm of cephalosporin C per minute at 25°C in the presence of EDTA pH 7.0.
Specifications
Specifications
7
4°C
1 Vial
32000
Lyophilized Solid
Destroy beta-Lactam antibiotics in body fluids or culture media and It is used in the production of Penicillin
Bacillus cereus
b-Lactamase
Clavulanic Acid, Sulbactam, Tazobactam