missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Albumin, Bovine 30%, Cap Free Sterile

Sterile

Brand:  MP Biomedicals™ 08810133

319.22 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.



Additional Details : CAS : 9048-46-8 Weight : 0.05000kg

Product Code. 11464216

  • 336.02 € / 50mL
Estimated Shipment: 19-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Albumin, Bovine, [9048-46-8]

  • F.W. 66338
  • 30% Solution
  • Cap Free

Specifications

Specifications

Immunoassay, Mammalian Cell Culture
Bovine Albumin
50mL
2° to 8°C
66338
7.4
Bovine Blood
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions