missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ki67/MKI67 Antibody, R&D Systems™
Mouse Monoclonal Antibody
248.00 € - 560.00 €
Specifications
| Antigen | Ki67/MKI67 |
|---|---|
| Clone | 1088403 |
| Dilution | Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279892
|
R&D Systems
MAB11612-SP |
25 μg |
248.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279911
|
R&D Systems
MAB11612-100 |
100 μg |
560.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ki67/MKI67 Monoclonal antibody specifically detects Ki67/MKI67 in Mouse samples. It is validated for Immunohistochemistry,ImmunocytochemistrySpecifications
| Ki67/MKI67 | |
| Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 4288 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
| 1088403 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Mouse | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126 | |
| Synthetic Peptide | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title