missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2c Rabbit anti-Mouse, Clone: RM223, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB15110.100ug
Additional Details : Weight : 0.00010kg
Description
Rabbit monoclonal antibody raised against native mouse IgG2c.
- High specificity
- High reactivity
Specifications
Mouse IgG2c | |
Monoclonal | |
Unconjugated | |
ELISA (0.01-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) Immunoprecipitation Western Blot (nonreduced only) (0.5-2 ug/mL) The optimal worki | |
IGHG2C | |
Rabbit | |
Protein A affinity purification | |
RUO | |
404711 | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
RM223 | |
Rabbit monoclonal antibody raised against native mouse IgG2c. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
Igh-1b | |
Native purified IgG2c from mouse hybridoma. | |
100 μg | |
Primary | |
This antibody reacts to the Fc region of mouse IgG2c. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG3, IgM, IgA, IgE, human IgG, or rat IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
For Research Use Only (RUO)