Learn More
FBXO5, Mouse, Clone: 5H7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FBXO5.
Brand: Abnova H00026271-M01A.200uL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is similar to xenopus early mitotic inhibitor-1 (Emi1), which is a mitotic regulator that interacts with Cdc20 and inhibits the anaphase promoting complex. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: RHNEFSEVAKTLKKNESLKACIRCNSPAKYDCYLQRATCKREGCGFDYCTKCLCNYHTTKDCSDGKLLKASCKIGPLPGTKKSKKNLRRSpecifications
FBXO5 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_012177 | |
FBXO5 | |
FBXO5 (NP_036309,358 a.a. ∽ 446 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
26271 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
5H7 | |
Mouse monoclonal antibody raised against a partial recombinant FBXO5. | |
FBXO5 | |
EMI1/FBX5/Fbxo31 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |