missing translation for 'onlineSavingsMsg'
Learn More

MOPS-buffered saline (5X), pH 7.5, Thermo Scientific™

Catalog Number p-7042186
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 5X
pH 7.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material MOPS-buffered saline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'