Learn More
Molybdenum AAS Standard, 1000 mg/L Mo in HNO3 1% HF 1%, Honeywell Fluka™
Brand:  Honeywell Fluka E4046-500ML
Additional Details : Weight : 0.50500kg ??product.attributeSet.Transport?? : UN number : 2922 Chem class : 8 Pack group : II
 View more versions of this product
Product Code. 16654344
  • 42.53 € / 500mL
Estimated Shipment: 24-01-2022
Add to basket
Description
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Specifications
Molybdenum AAS Standard
1.01 g/cm3
<2
500 mL