missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Magnesium Green™, AM, cell permeant - Special Packaging

Exhibits higher affinity for Mg2+ (Kd approximately 1.0mM) than mag-fura-2 (Kd approximately 1.9mM) or mag-indo-1 (Kd approximately 2.7mM)

Brand:  Invitrogen™ M3735

498.69 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.00450kg

Product Code. 11538966

  • 748.00 € / Pack of 20
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

 

  • Upon binding Mg2+, Magnesium Green exhibits an increase in fluorescence emission intensity without shift in wavelength
  • Absorption/emission maxima approximately 506/531nm

 

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Magnesium Detection

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer -5°C to -30°C.
Cell Viability and Proliferation
Magnesium Indicator
Magnesium Green™
Fluorescent
Fluorescence Microscope, Microplate Reader
Magnesium Indicator
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.