missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 233

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 233 is a non-peptide apelin receptor agonist (EC50 = 3.7 μM); exhibits >21-fold selectivity over the closely related angiotensin 1 receptor (EC50 = >79 μM).
Specifications
Specifications
| Chemical Name or Material | (E)-4-Methyl-5-(((phenylsulfonyl)oxy)imino)-[1,1'-bi(cyclohexane)]-3,6-dien-2-one |
| CAS | 143621-35-6 |
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in DMSO |
| Molecular Formula | C19H21NO4S |
| Formula Weight | 359.44 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction