Learn More
Methyl Ester of Cetirizine Dihydrochloride, Mikromol™
Product Code. 30050487
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30050487
Brand: Mikromol MM0380.160025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl Ester of Cetirizine Dihydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl Ester of Cetirizine Dihydrochloride
CAS 83881-48-5
Analyte or Component Names Methyl Ester of Cetirizine Dihydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H27 Cl N2 O3 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Methyl Ester of Cetirizine Dihydrochloride
InChI Formula InChI=1S/C22H27ClN2O3.2ClH/c1-27-21(26)17-28-16-15-24-11-13-25(14-12-24)22(18-5-3-2-4-6-18)19-7-9-20(23)10-8-19;;/h2-10,22H,11-17H2,1H3;2*1H
SMILES Cl.Cl.COC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name methyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate;dihydrochloride
Molecular Weight (g/mol) 475.84
Quantity 25 mg
Formula Weight 474.1244 g/mol