missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.
Solubility
Soluble in water. Insoluble in ether
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.
Solubility
Soluble in water. Insoluble in ether
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
Specifications
Melting Point | 166°C to 171°C |
Density | 1.46 |
Quantity | 100 g |
Beilstein | 81568 |
Sensitivity | Hygroscopic |
Merck Index | 14,6080 |
Solubility Information | Soluble in water; Insoluble in ether |
Optical Rotation | +159° (c=10 in Water) |
Formula Weight | 194.18 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'