Learn More
Methyl Acetate, ReagentPlus™, 99%, Honeywell™
Catalog Number p-7120580
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
79-20-9
C3H6O2
74.079
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
79-20-9
C3H6O2
74.079
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
Reagent Grade, 99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -98°C
Color Colorless
Density 0.934g/mL at 25°C
Boiling Point 58°C
Flash Point −13°C (8.6°F)
Packaging Glass Bottle
Linear Formula CH3COOCH3
Refractive Index n20/D 1.361(lit.)
Quantity 2.5 L
UN Number UN1231
Beilstein 1736662
Vapor Density 2.55 (vs air)
Formula Weight 74.08g/mol
Percent Purity 99%
Grade ReagentPlus™
Physical Form Liquid
Chemical Name or Material Methyl Acetate