Learn More
Methyl 2-(Bromomethyl)-6-nitrobenzoate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Methyl 2-(Bromomethyl)-6-nitrobenzoate
Specifications
Specifications
| Chemical Name or Material | 2-Bromomethyl-6-nitrobenzoic acid methyl ester |
| CAS | 61940-21-4 |
| Analyte or Component Names | 2-Bromomethyl-6-nitrobenzoic acid methyl ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H8 Br N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzoic acid, 2-(bromomethyl)-6-nitro-, methyl ester (9CI, ACI), Methyl 2-(2-bromomethyl)-6-nitrobenzoate, Methyl 2-(Bromomethyl)-6-nitrobenzoate |
| InChI Formula | InChI=1S/C9H8BrNO4/c1-15-9(12)8-6(5-10)3-2-4-7(8)11(13)14/h2-4H,5H2,1H3 |
| SMILES | COC(=O)c1c(CBr)cccc1[N+](=O)[O-] |
| IUPAC Name | methyl 2-(bromomethyl)-6-nitrobenzoate |
| Show More |