missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricyclazole in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Specifications
Concentration | 100μg/mL |
Packaging | Ampoule |
Quantity | 1 mL |
Analyte or Component Names | Tricyclazole in Methanol |
Solvent or Matrix | Methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'