missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropionic acid methyl ester in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™

Catalog Number 12996043
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 1000μg/mL
CAS 1729-67-5
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C4H6Br2O2
InChI Key ROXQOUUAPQUMLN-UHFFFAOYSA-N
SMILES COC(=O)C(CBr)Br
IUPAC Name methyl 2,3-dibromopropanoate
Molecular Weight (g/mol) 245.898
PubChem CID 95428
Solvent or Matrix Methyl-tert.butylether
Chemical Name or Material 2,3-Dibromopropionic acid methyl ester 
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'