missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 5.5
Click to view available options
Quantity:
250 mL
500 mL
Description
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Color | Colorless |
Concentration | 0.2 M |
pH | 5.5 |
Physical Form | Liquid |
Quantity | 250 mL |
Solubility Information | Fully miscible. |
Chemical Name or Material | MES buffer soln. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'