missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MELK Recombinant Human Protein

MELK is a serine/threonione kinase which is related to the AMPK family.

1826.00 € - 7890.00 €

Specifications

Accession Number NP_055606.1
Formulation 50mM Tris, pH 7.5, 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT
Molecular Weight (g/mol) 66.8kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Regulatory Status RUO - research use only
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418703
View Documents Promotion Details
Invitrogen™
PR8926B
100μg
1826.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15468763
View Documents Promotion Details
Invitrogen™
PV4824
1mg
7890.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MELK has been linked to the cell cycle. It has been shown to be overexpressed in cancer and has been identified has a potential target for cancer therapeutics.

Specifications

Specifications

NP_055606.1
66.8kDa
RUO - research use only
MELK
Human
GST Tag
Frozen
50mM Tris, pH 7.5, 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
CAMKL Kinase Family
Recombinant
Baculovirus
Videos
SDS
Documents

Documents

Product Certifications