missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Medium 154CF/PRF, Kit
Medium 154CF/PRF, Kit
Brand: Gibco™ M154CFPRF500
Additional Details : Weight : 0.45000kg
Description
A sterile, liquid medium for the culture of human epidermal keratinocytes. Medium 154CF⁄PRF is Medium 154 prepared without calcium chloride or phenol red. Calcium chloride is provided as a separate component with each bottle of medium. This basal medium requires the addition of HKGS (S-001-5) or HKGS Kit (S-001-K) prior to use.Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Serum-free | |
Liquid | |
Serum-free | |
Sterile-filtered | |
Sustainable packaging | |
Room Temperature |
Keratinocyte Cells | |
Mammalian Cell Culture | |
Medium 154CF Kit | |
Sterile-filtered | |
No Phenol Red | |
Cascade Biologics™ |
For Research Use Only. Not for use in diagnostic procedures.