Learn More
MCC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557933
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VQELVSALER LTKSSEIRHQ QSAEFVNDLK RANSNLVAAY EKAKKKHQNK LKKLESQMMA MVERHETQVR MLKQRIALLE EENSRPHTNE TS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
MCC Polyclonal antibody specifically detects MCC in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MCC | |
Polyclonal | |
Unconjugated | |
Mcc | |
colorectal mutant cancer protein; D18Ertd451e; E330037C19; MCC; MCC1; mutated in colorectal cancers; Protein MCC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23508 | |
Mcc | |
Recombinant protein corresponding to Human MCC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only