missing translation for 'onlineSavingsMsg'
Learn More

Amarell™ Maximum Thermometer with Steel Mark

A fully immersible thermometer for quick and accurate temperature readings.

Brand:  Amarell™ G13012

Additional Details : Weight : 0.04300kg

Product Code. 10463721

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Fully immersible
  • Enclosed scale
  • Capillary prismatic
  • Mercury filling
  • Not to be exposed to ambient temperatures of 200°C or more

Specifications

Specifications

Total Immersion
300 mm
Mercury
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions