Learn More
Abnova™ MaxBead Protein A
MaxBead Protein A is magnetic bead designed for capturing of IgG or antigen via protein A-IgG-antigen interaction.
Brand:  Abnova™ U0084.10x1mL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16072378
  • 648.63 € / Pack of 10
Estimated Shipment: 14-12-2021
Add to basket
Description
Description
Specifications
Specifications
Purification
10 x 1 mL