missing translation for 'onlineSavingsMsg'
Learn More

Manganous Sulfate, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390789
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
10034-96-5
H2MnO5S
169.01
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10034-96-5
H2MnO5S
169.01
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 500 g
IUPAC Name manganese(2+);sulfate;hydrate
Formula Weight 169.02
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.95kg
Chemical Name or Material Manganous Sulfate, Monohydrate
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'