Learn More
Invitrogen™ MagniSort™ Human pan-Monocyte Enrichment Kit
Designed for the magnetic separation of conventional CD14+ and unconventional CD16+CD14- monocytes by negative selection.
Brand: Invitrogen™ 8804-6837-74
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes: MagniSort Human pan-Monocyte Enrichment Antibody Cocktail : 200 tests, 20μL/test
MagniSort Negative Selection Beads B
Description
- Undesired cells, including CD16+ NK cells, are bound by antibody and then magnetic beads
- When placed in a magnetic field, monocytes are untouched and free in solution
The MagniSort™ Human pan-Monocyte Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD2 Biotin
Anti-Human CD3 Biotin
Anti-Human CD7 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD56 Biotin
Anti-Human CD57 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Specifications
Monocytes (pan) | |
Human Pan-Monocyte Enrichment Kit | |
Human |
For Research Use Only. Not for use in diagnostic procedures.