missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, ≥99%, Reagent Grade, Honeywell™

Catalog Number p-7120528
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
16674-78-5
C4H14MgO8
214.45
MFCD00149214
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
134717
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
16674-78-5
C4H14MgO8
214.45
MFCD00149214
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
134717
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point 68°C
Color Colorless
pH 8 to 9 (at 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula (CH3COO)2Mg · 4H2O
UN Number NONH for all modes of transport
Beilstein 3730605
Solubility Information Completely soluble in water
IUPAC Name magnesium(2+) diacetate tetrahydrate
Formula Weight 214.45g/mol
Percent Purity ≥99%
Odor Vinegar-like
Packaging Poly bottle
Density 1.45g/cu.cm at 20°C
Chemical Name or Material Magnesium acetate tetrahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'