missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LP 922056
GREENER_CHOICE

Product Code. 18759946
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18759946

Brand: Tocris Bioscience™ 6691/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Notum inhibitor; activates Wnt signaling

LP 922056 is a notum pectinacetylesterase inhibitor. LP 922056 activates wnt signaling in vitro (EC50 values are 21 nM in human and 55 nM in mouse cell-based assay). It increases cortical bone thickness in mouse model of bone growth. LP 922056 also inhibits growth of Apc-mutant cell lines and proliferation of intestinal adenomas in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(6-Chloro-7-cyclopropylthieno[3,2-d]pyrimidin-4-yl)thio]acetic acid
CAS 1365060-22-5
Quantity 50 mg
Molecular Formula C11H9ClN2O2S2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.