Learn More
Lovibond™ Aqua Comparitor Color Discs: Zinc
For testing water with the Aqua Comparator
Brand: Lovibond™ 146340
26.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for zinc using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0 to 1mg/L | |
Zinc LR Color Disc |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.