missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Lithium trifluoromethanesulfonate is mixed with ceramic filler to prepare polyethylene oxide films in order to improve the electrochemical and mechanical characteristics of the membrane, which is used in the lithium batteries. It acts as a doping salt used in the preparation of nano manganese-composite polymer electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 423°C |
Color | White |
Quantity | 10 g |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with water. |
Formula Weight | 156.01 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | Lithium trifluoromethanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'