Learn More
KNF Neuberger™ Liquiport™ Analog Diaphragm Liquid Pump
LIQUIPORT™ pumps offer a multipurpose pump ideal for aspiration or transferring liquids. Flows can be adjusted manually or by motor speed controlled with RC versions.
Brand:  KNF Neuberger™ 153690/153257
Additional Details : Weight : 1.50000kg
Product Code. 10143992
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Self priming
  • Can be run dry
  • Minimal maintenance and long life
  • IP65 motor protection class
  • Outstanding chemical resistance (KT versions)
  • Quiet operation