Learn More
KNF Neuberger™ Liquiport™ Analog Diaphragm Liquid Pump
LIQUIPORT™ pumps offer a multipurpose pump ideal for aspiration or transferring liquids. Flows can be adjusted manually or by motor speed controlled with RC versions.
Brand: KNF Neuberger™ 153690/153257
Additional Details : Weight : 1.50000kg
Description
- Self priming
- Can be run dry
- Minimal maintenance and long life
- IP65 motor protection class
- Outstanding chemical resistance (KT versions)
- Quiet operation