Learn More
Lactam of (S)-Carboxymethyl-L-cysteine, Mikromol™
Product Code. 16324396
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 16324396
Brand: Mikromol MM0236.02
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Lactam of (S)-Carboxymethyl-L-cysteine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Lactam of (S)-Carboxymethyl-L-cysteine
CAS 62305-89-9
Analyte or Component Names Lactam of (S)-Carboxymethyl-L-cysteine
Shipping Condition Room Temperature
Molecular Formula C5 H7 N O3 S
Certifications/Compliance ISO 17025
Synonym Lactam of (S)-Carboxymethyl-L-cysteine, 3-Thiomorpholinecarboxylic acid, 5-oxo-, (3R)-, 3-Thiomorpholinecarboxylic acid, 5-oxo-, (R)-, (3R)-3-Carboxy-5-oxothiomorpholine
InChI Formula InChI=1S/C5H7NO3S/c7-4-2-10-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1
SMILES OC(=O)[C@@H]1CSCC(=O)N1
IUPAC Name (3R)-5-oxothiomorpholine-3-carboxylic acid
Molecular Weight (g/mol) 161.179
Quantity 100 mg
Formula Weight 161.0147 g/mol