Learn More
Deutsch Neumann™ Laboratory Tubing
Provides flexibility, tear strength and resilience for general laboratory usage. Deutsch Neumann™ Laboratory Tubing is manufactured from red natural rubber.
Brand:  Deutsch Neumann™ 3010609
Additional Details : Weight : 1.35000kg
 View more versions of this product
Product Code. 10048320
Quantity Price
1 1.90 € / 1m
25 1.60 € / 1m
In stock, ready to ship: 14
Add to basket
Description
Description
  • Durometer 50 Shore A
  • Good elastic memory
Specifications
Especificaciones
ISO 9001:2008
6 mm
9 mm
1.5 mm
Red
Natural Rubber
25 m
For Laboratory Use
Videos
SDS
Documentos
Certificados