missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine benzyl ester p-toluenesulfonate, 98%, Thermo Scientific™

Catalog Number p-7041529
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1738-77-8
C20H27NO5S
393.50
MFCD00066117
QTQGHKVYLQBJLO-YDALLXLXSA-N
l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh
6451312
benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
1738-77-8
C20H27NO5S
393.50
MFCD00066117
QTQGHKVYLQBJLO-YDALLXLXSA-N
l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh
6451312
benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 160°C
Beilstein 3566840
Formula Weight 393.5
Percent Purity 98%
Quantity 1 g
Chemical Name or Material L-Leucine benzyl ester p-toluenesulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'