missing translation for 'onlineSavingsMsg'
Learn More

L-Cyclopentylglycine, 95%, Thermo Scientific Chemicals

Catalog Number p-7041602
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2521-84-8
C7H13NO2
143.186
MFCD03788076
XBPKRVHTESHFAA-LURJTMIESA-N
l-cyclopentylglycine, s-2-amino-2-cyclopentylacetic acid, 2-cyclopentyl-l-glycine, h-cyclopentyl-gly-oh, l-beta-cyclopentylglycine, 2s-amino cyclopentyl ethanoic acid, h-l-cpg-oh, s-2-cyclopentylglycine, s-amino-cyclopentyl-acetic acid, 2s-2-amino-2-cyclopentylacetic acid
10374564
(2S)-2-amino-2-cyclopentylacetic acid
C1CCC(C1)C(C(=O)O)N
This item is not returnable. View return policy
2521-84-8
C7H13NO2
143.186
MFCD03788076
XBPKRVHTESHFAA-LURJTMIESA-N
l-cyclopentylglycine, s-2-amino-2-cyclopentylacetic acid, 2-cyclopentyl-l-glycine, h-cyclopentyl-gly-oh, l-beta-cyclopentylglycine, 2s-amino cyclopentyl ethanoic acid, h-l-cpg-oh, s-2-cyclopentylglycine, s-amino-cyclopentyl-acetic acid, 2s-2-amino-2-cyclopentylacetic acid
10374564
(2S)-2-amino-2-cyclopentylacetic acid
C1CCC(C1)C(C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 143.18
Percent Purity 95%
Quantity 250 mg
Chemical Name or Material L-Cyclopentylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'