missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off button top protects target area and allows easy access
Brand: DWK Life Sciences 73843B-13
Additional Details : Weight : 0.10000kg
Description
- For use with 13 and 20mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
- Allows easy access with a flip of the thumb
- Available with blue, red, green and white plastic buttons to help separate and identify samples
Specifications
Blue | |
Autoclavable | |
Flip-up Button-top Cap |
Aluminum | |
1000 Cs. | |
13 mm |