missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Brand Colorware Volumetric Class A Flasks

Single graduation ring blasted on the neck and durable matte finish marking area for use with an ordinary pencil

Brand:  DWK Life Sciences 28014Y-25

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 10039761

  • 323.00 € / Pack of 6

Estimated Shipment: 21-06-2022
to see stock.
Add to basket

Description

Description

  • Calibrated to contain with Class A accuracy but without serial number
  • Standard taper ground glass stopper is provided
  • Designed from ASTM Specification E288, Class A unserialized requirements

Specifications

Specifications

Glass
25 mL
4.64 in.
±0.03 mL
9
Sunny Yellow
Borosilicate Glass
11.8 cm
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions