missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric anhydride, 95%, Thermo Scientific Chemicals

Catalog Number p-4813500
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1468-39-9
C10H18O3
186.25
MFCD00015051
FREZLSIGWNCSOQ-UHFFFAOYSA-N
isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
73847
3-methylbutanoyl 3-methylbutanoate
CC(C)CC(=O)OC(=O)CC(C)C
This item is not returnable. View return policy
1468-39-9
C10H18O3
186.25
MFCD00015051
FREZLSIGWNCSOQ-UHFFFAOYSA-N
isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
73847
3-methylbutanoyl 3-methylbutanoate
CC(C)CC(=O)OC(=O)CC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9400g/mL
Boiling Point 215°C
Flash Point 91°C
Assay Percent Range 94% min
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 0.94
Solubility Information Solubility in water: hydrolyses
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material Isovaleric anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Isovaleric anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'