Learn More
Isoindoline hydrochloride, 97%, Thermo Scientific Chemicals
Catalog Number p-4228752
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
32372-82-0
C8H10ClN
155.63
MFCD03502377
NOVIRODZMIZUPA-UHFFFAOYSA-N
isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
12311031
Cl.C1NCC2=CC=CC=C12
32372-82-0
C8H10ClN
155.63
MFCD03502377
NOVIRODZMIZUPA-UHFFFAOYSA-N
isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
12311031
Cl.C1NCC2=CC=CC=C12
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 255°C to 256°C
Color White
Elemental Analysis 59.27 to 64.21% C, 8.64 to 9.36% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name 2,3-dihydro-1H-isoindole hydrochloride
Formula Weight 155.63
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Isoindoline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Isoindoline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.