missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Chloride Hexahydrate, Carl Roth™

Catalog Number 10681393
Encompass
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy
Melting Point 37°C
Color Brown-Yellow
pH ∼1.8
Physical Form Solid
Quantity 1 kg
Concentration or Composition (by Analyte or Components) 270.33g/mol
Odor Chlorine
Density 1.82g/cm3
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'