Learn More
IKA™ IO T 1.10 Grid Tray
Enhanced wire grid tray designed to be used with an IKA™ Drying Oven.
Brand:  IKA™ 0020007600
215.38 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 2.84000kg
Product Code. 16208461
  • 275.00 € / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • For use with the following IKA™ Drying Ovens:
    • Oven 125 Basic Dry
    • Oven 125 Basic Dry, Glass
    • Oven 125 Control Dry
    • Oven 125 Control Dry, Glass
Specifications
Specifications
2.84 kg