Learn More
Invitrogen™ Qubit™ dsDNA HS Assay Kit
Highly selective for double-stranded DNA (dsDNA) over RNA
103.00 € - 304.00 €
Benötigt: Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes
Produkte 2
Produktcode Marke Menge. Preis Menge & Verfügbarkeit  
Produktcode Marke Menge. Preis Menge & Verfügbarkeit  
10616763 Invitrogen™
Q32854
500 assays 304.00 €
1 Set Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Auf Lager: 250
Zum Warenkorb hinzufügen
10606433 Invitrogen™
Q32851
100 assays 103.00 €
100 Assays Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Auf Lager: 152
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The Qubit dsDNA HS (High Sensitivity) Assay Kit is designed specifically for use with the Qubit Fluorometer. The assay is highly selective for double-stranded DNA (dsDNA) over RNA and is designed to be accurate for initial sample concentrations from 10 pg/µL to 100 ng/µL. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 µl and 20 µl is acceptable), and read the concentration using the Qubit Fluorometer. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein are well tolerated in the assay.
Which product to choose for dsDNA HS (High Sensitivity) quantitation?
• For 1–20 samples: use this Qubit dsDNA HS Assay Kit with the Qubit Fluorometer
• For 20–2000 samples: use the Quant-iT dsDNA HS Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500 µL thin-walled PCR tubes are required but not included.
3. Also available as a "1X" dsDNA assay kit that provides reagent and buffer in a formulation that is stable as a ready-to-use solution for up to one year after preparation.
Order Info
Shipping Condition: Room temperature
For Research Use Only. Not for use in diagnostic procedures.