missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Insulating Cooling Pack
Brand: Carl Roth™ CY97.1
Additional Details : Weight : 0.10000kg
Specifications
450 x 200 mm | |
Insulating cooling pack in flexible sleeve | |
Microfiber |
Cooling and keeping cool of bottles with approx. 70 to 100 mm diameter | |
200 mm | |
450 mm |