Learn More
B Braun™ Needle-Pro™ Hypodermic Needle
B Braun™ Needle-Pro™ Hypodermic Needle is specially designed for protection from accidental needle sticks.
Brand: B Braun™ 4658300
Additional Details : Weight : 0.50000kg
Description
- Protects from accidental needle sticks
- Simple, intuitive handling
- Sensible, audible activation of security mechanism
- Permanent enclosure of the cannula by a stable, protective cover
- Uncomplicated disposal in suitable cannula disposal containers
- Latex-free
Specifications
Gray | |
27 G | |
12 mm | |
100/Pk., 800/Cs. |