Learn More
IL-2 Monoclonal Antibody (MQ1-17H12), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
130.00 € - 311.00 €
Specifications
Antigen | IL-2 |
---|---|
Clone | MQ1-17H12 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15370820
|
Affymetrix eBioscience
17-7029-82 |
100 μg |
311.00 €
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15528186
|
Affymetrix eBioscience
17-7029-41 |
25 Tests |
130.00 €
25 tests |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody. Applications Reported: MQ1-17H12 has been reported for use in staining of intracellular IL-2. Applications Tested: This MQ1-17H12 antibody is tested by intracellular staining and flow cytometric analysis of activated human normal human peripheral blood cells.This product has been pre-titrated and tested intracellular staining and flow cytometric analysis of activated human normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Flow Cytometry.Specifications
IL-2 | |
Flow Cytometry | |
APC | |
Rat | |
Human | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
IL2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2 |
MQ1-17H12 | |
Monoclonal | |
Liquid | |
RUO | |
P60568 | |
3558 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.