Learn More
Rat IgG2a kappa Isotype Control (eBR2a), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 45-4321-80
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG2a kappa is useful as an isotype control immunoglobulin. Applications Reported: PerCP-Cy5.5 Rat IgG2a Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2a Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2a kappa | |
eBR2a | |
PerCP-Cyanine5.5 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a κ |
For Research Use Only.